Assay Details
Gene Symbol: acsl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003569.2 | NP_001003569.1 | 5 - 6 | 602 | 98 | |
XM_005169565.4 | XP_005169622.1 | 5 - 6 | 652 | 98 | ||
GenBank mRNA | BC077120.1 | - | 5 - 6 | 603 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
