Assay Details
Gene Symbol: mrpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017696.1 | NP_001017696.1 | 3 - 4 | 478 | 127 | |
GenBank mRNA | BC093312.1 | - | 3 - 4 | 478 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
