Assay Details
Gene Symbol: tprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030123.1 | NP_001025294.1 | 11 - 12 | 1204 | 145 | |
XM_009294751.3 | XP_009293026.1 | 11 - 12 | 1225 | 145 | ||
GenBank mRNA | BC134977.1 | - | 11 - 12 | 1202 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
