Assay Details
Gene Symbol: bud23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082879.1 | NP_001076348.1 | 7 - 8 | 616 | 70 | |
GenBank mRNA | BC133941.1 | - | 7 - 8 | 616 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
