Assay Details
Gene Symbol: myo1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100031.1 | NP_001093501.1 | 13 - 14 | 1544 | 112 | |
XM_005165347.4 | XP_005165404.1 | 13 - 14 | 1719 | 112 | ||
XM_005165349.4 | XP_005165406.1 | 13 - 14 | 1779 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
