Assay Details
Gene Symbol: ss18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291325.1 | NP_001278254.1 | 3 - 4 | 357 | 84 | |
NM_199744.2 | NP_956038.1 | 3 - 4 | 357 | 84 | ||
GenBank mRNA | BC047188.1 | - | 3 - 4 | 333 | 84 | |
EH553447.1 | - | 3 - 4 | 397 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
