Assay Details
Gene Symbol: ss18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291325.1 | NP_001278254.1 | 6 - 7 | 869 | 91 | |
NM_199744.2 | NP_956038.1 | 6 - 7 | 869 | 91 | ||
GenBank mRNA | BC047188.1 | - | 6 - 7 | 845 | 91 | |
EH561714.1 | - | 4 - 5 | 637 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
