Assay Details
Gene Symbol: ndufb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200855.1 | NP_957149.1 | 2 - 3 | 228 | 98 | |
GenBank mRNA | BC062842.1 | - | 2 - 3 | 228 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
