Assay Details
Gene Symbol: arhgdig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201157.1 | NP_957451.1 | 3 - 4 | 282 | 107 | |
GenBank mRNA | BC056296.1 | - | 3 - 4 | 282 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
