Assay Details
Gene Symbol: pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213387.1 | NP_998552.1 | 2 - 3 | 186 | 139 | |
GenBank mRNA | BC046026.1 | - | 2 - 3 | 186 | 139 | |
BC065888.1 | - | 2 - 3 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
