Assay Details
Gene Symbol: ociad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044856.2 | NP_001038321.1 | 6 - 7 | 532 | 70 | |
GenBank mRNA | BC095762.1 | - | 6 - 7 | 532 | 70 | |
BC134089.1 | - | 6 - 7 | 532 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
