Assay Details
Gene Symbol: bmi1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194366.1 | NP_919347.1 | 4 - 5 | 615 | 76 | |
GenBank mRNA | AB074153.1 | - | 4 - 5 | 615 | 76 | |
BC049423.1 | - | 4 - 5 | 375 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
