Assay Details
Gene Symbol: bmi1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194366.1 | NP_919347.1 | 5 - 6 | 666 | 111 | |
GenBank mRNA | AB074153.1 | - | 5 - 6 | 666 | 111 | |
BC049423.1 | - | 5 - 6 | 426 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
