Assay Details
Gene Symbol: rassf7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197942.1 | NP_922933.1 | 3 - 4 | 1155 | 66 | |
XM_005166462.4 | XP_005166519.1 | 3 - 4 | 1255 | 66 | ||
XM_005166464.4 | XP_005166521.1 | 3 - 4 | 1152 | 66 | ||
XM_017356819.2 | XP_017212308.1 | 3 - 4 | 1253 | 66 | ||
GenBank mRNA | BC044177.1 | - | 3 - 4 | 1155 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
