Assay Details
Gene Symbol: rassf7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197942.1 | NP_922933.1 | 5 - 6 | 1419 | 106 | |
XM_005166462.4 | XP_005166519.1 | 5 - 6 | 1519 | 106 | ||
XM_005166464.4 | XP_005166521.1 | 5 - 6 | 1416 | 106 | ||
XM_017356819.2 | XP_017212308.1 | 5 - 6 | 1517 | 106 | ||
GenBank mRNA | BC044177.1 | - | 5 - 6 | 1419 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
