Assay Details
Gene Symbol: vti1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201036.1 | NP_957330.1 | 5 - 6 | 714 | 67 | |
GenBank mRNA | BC055131.1 | - | 5 - 6 | 714 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
