Assay Details
Gene Symbol: rps21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201191.1 | NP_957485.1 | 5 - 6 | 314 | 72 | |
GenBank mRNA | BC049056.1 | - | 5 - 6 | 314 | 72 | |
BC071475.1 | - | 5 - 6 | 314 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
