Assay Details
Gene Symbol: tnfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212859.2 | NP_998024.2 | 3 - 4 | 375 | 70 | |
GenBank mRNA | AY427649.1 | - | 3 - 4 | 278 | 70 | |
BC124141.1 | - | 3 - 4 | 375 | 70 | ||
BC165066.1 | - | 3 - 4 | 312 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
