Assay Details
Gene Symbol: pik3r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291329.1 | NP_001278258.1 | 7 - 8 | 965 | 63 | |
NM_213038.2 | NP_998203.2 | 6 - 7 | 1128 | 63 | ||
GenBank mRNA | BC059572.1 | - | 6 - 7 | 1109 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
