Assay Details
Gene Symbol: cfl1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213639.2 | NP_998804.1 | 2 - 3 | 644 | 72 | |
GenBank mRNA | AY398323.1 | - | 2 - 3 | 472 | 72 | |
BC049463.1 | - | 2 - 3 | 391 | 72 | ||
EH548817.1 | - | 2 - 3 | 666 | 72 | ||
EH574450.1 | - | 2 - 3 | 499 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
