Assay Details
Gene Symbol: cfl1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213639.2 | NP_998804.1 | 3 - 4 | 712 | 133 | |
GenBank mRNA | AY398323.1 | - | 3 - 4 | 540 | 133 | |
BC049463.1 | - | 3 - 4 | 459 | 133 | ||
EH548817.1 | - | 3 - 4 | 734 | 133 | ||
EH574450.1 | - | 1 - 2 | 431 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
