Assay Details
Gene Symbol: sap30l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214703.1 | NP_999868.1 | 2 - 3 | 374 | 77 | |
GenBank mRNA | BC066447.1 | - | 2 - 3 | 374 | 77 |
Gene Symbol: LOC103910016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009298403.3 | XP_009296678.3 | 2 - 3 | 477 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
