Assay Details
Gene Symbol: tbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214796.1 | NP_999961.1 | 4 - 5 | 916 | 120 | |
GenBank mRNA | AY256970.1 | - | 4 - 5 | 916 | 120 | |
BC116553.1 | - | 4 - 5 | 743 | 120 | ||
BC164097.1 | - | 4 - 5 | 633 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
