Assay Details
Gene Symbol: baxb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013296.2 | NP_001013314.1 | 4 - 5 | 461 | 163 | |
GenBank mRNA | BC090280.1 | - | 4 - 5 | 465 | 163 | |
DQ860150.1 | - | 4 - 5 | 465 | 163 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
