Assay Details
Gene Symbol: gria3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198339.1 | NP_938153.1 | 8 - 9 | 1220 | 126 | |
XM_005165095.4 | XP_005165152.1 | 8 - 9 | 1686 | 126 | ||
XM_005165096.4 | XP_005165153.1 | 8 - 9 | 1680 | 126 | ||
XM_005165097.4 | XP_005165154.1 | 8 - 9 | 1682 | 126 | ||
XM_005165098.4 | XP_005165155.1 | 8 - 9 | 1684 | 126 | ||
XM_021475991.1 | XP_021331666.1 | 8 - 9 | 1684 | 126 | ||
GenBank mRNA | AF525745.1 | - | 8 - 9 | 1220 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
