Assay Details
Gene Symbol: taf5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199852.1 | NP_956146.1 | 6 - 7 | 1130 | 116 | |
GenBank mRNA | BC056820.1 | - | 6 - 7 | 1130 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
