Assay Details
Gene Symbol: mycb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200172.1 | NP_956466.1 | 2 - 3 | 818 | 77 | |
GenBank mRNA | BC045424.1 | - | 2 - 3 | 818 | 77 | |
BC164674.1 | - | 1 - 2 | 680 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
