Assay Details
Gene Symbol: vamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213398.1 | NP_998563.1 | 4 - 5 | 371 | 127 | |
XM_005159424.4 | XP_005159481.1 | 5 - 6 | 697 | 127 | ||
GenBank mRNA | BC059687.1 | - | 4 - 5 | 371 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
