Assay Details
Gene Symbol: acaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002207.1 | NP_001002207.1 | 1 - 2 | 162 | 86 | |
GenBank mRNA | BC072706.1 | - | 1 - 2 | 162 | 86 | |
BC165883.1 | - | 1 - 2 | 191 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
