Assay Details
Gene Symbol: arl6ip5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002517.2 | NP_001002517.1 | 2 - 3 | 417 | 68 | |
GenBank mRNA | BC076281.1 | - | 2 - 3 | 418 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
