Assay Details
Gene Symbol: tbc1d16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006076.2 | NP_001006076.2 | 3 - 4 | 715 | 59 | |
GenBank mRNA | BC083244.1 | - | 3 - 4 | 713 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
