Assay Details
Gene Symbol: rras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017815.1 | NP_001017815.1 | 5 - 6 | 757 | 76 | |
GenBank mRNA | BC092803.1 | - | 5 - 6 | 757 | 76 | |
BC165269.1 | - | 5 - 6 | 552 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
