Assay Details
Gene Symbol: il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045279.1 | NP_001038744.1 | 2 - 3 | 213 | 110 | |
GenBank mRNA | BC116595.1 | - | 2 - 3 | 213 | 110 | |
EF014959.1 | - | 2 - 3 | 234 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
