Assay Details
Gene Symbol: g6pc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080066.1 | NP_001073535.1 | 5 - 6 | 780 | 94 | |
GenBank mRNA | BC128866.1 | - | 5 - 6 | 780 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
