Assay Details
Gene Symbol: msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082893.2 | NP_001076362.1 | 4 - 5 | 427 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
