Assay Details
Gene Symbol: rnf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131213.2 | NP_571288.2 | 2 - 3 | 155 | 77 | |
GenBank mRNA | AF196346.1 | - | 2 - 3 | 129 | 77 | |
AY576996.1 | - | 2 - 3 | 162 | 77 | ||
BC044472.1 | - | 2 - 3 | 152 | 77 | ||
BC164137.1 | - | 1 - 2 | 124 | 77 | ||
EH435872.1 | - | 2 - 3 | 167 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
