Assay Details
Gene Symbol: rnf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131213.2 | NP_571288.2 | 4 - 5 | 530 | 82 | |
GenBank mRNA | AF196346.1 | - | 4 - 5 | 504 | 82 | |
AY576996.1 | - | 4 - 5 | 537 | 82 | ||
BC044472.1 | - | 4 - 5 | 527 | 82 | ||
BC164137.1 | - | 3 - 4 | 499 | 82 | ||
EH435872.1 | - | 4 - 5 | 542 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
