Assay Details
Gene Symbol: rnf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131213.2 | NP_571288.2 | 6 - 7 | 972 | 90 | |
GenBank mRNA | BC044472.1 | - | 6 - 7 | 969 | 90 | |
BC164137.1 | - | 5 - 6 | 941 | 90 | ||
EH593592.1 | - | 1 - 2 | 181 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
