Assay Details
Gene Symbol: glra4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131782.2 | NP_571857.1 | 8 - 9 | 1583 | 65 | |
XM_005169598.4 | XP_005169655.1 | 8 - 9 | 1514 | 65 | ||
GenBank mRNA | AJ404970.1 | - | 8 - 9 | 1587 | 65 | |
BC090676.1 | - | 8 - 9 | 1238 | 65 | ||
GQ406229.1 | - | 8 - 9 | 1071 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
