Assay Details
Gene Symbol: ero1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200350.2 | NP_956644.2 | 3 - 4 | 463 | 63 | |
GenBank mRNA | BC053166.1 | - | 3 - 4 | 358 | 63 | |
BC165610.1 | - | 3 - 4 | 328 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
