Assay Details
Gene Symbol: stn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200389.1 | NP_956683.1 | 7 - 8 | 1012 | 133 | |
GenBank mRNA | BC053298.1 | - | 7 - 8 | 1012 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
