Assay Details
Gene Symbol: boka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003612.2 | NP_001003612.2 | 2 - 3 | 756 | 149 | |
GenBank mRNA | BC078217.1 | - | 2 - 3 | 719 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
