Assay Details
Gene Symbol: boka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003612.2 | NP_001003612.2 | 3 - 4 | 793 | 98 | |
GenBank mRNA | BC078217.1 | - | 3 - 4 | 756 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
