Assay Details
Gene Symbol: boka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003612.2 | NP_001003612.2 | 4 - 5 | 961 | 66 | |
XM_005165921.4 | XP_005165978.1 | 3 - 4 | 935 | 66 | ||
GenBank mRNA | BC078217.1 | - | 4 - 5 | 924 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
