Assay Details
Gene Symbol: spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077250.2 | NP_001070718.2 | 7 - 8 | 1162 | 71 | |
GenBank mRNA | BC115335.1 | - | 7 - 8 | 919 | 71 | |
BC124739.1 | - | 7 - 8 | 1122 | 71 | ||
EB952759.1 | - | 4 - 5 | 453 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
