Assay Details
Gene Symbol: tnfrsfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131840.2 | NP_571915.2 | 2 - 3 | 421 | 88 | |
GenBank mRNA | AF250042.2 | - | 2 - 3 | 351 | 88 | |
BC162652.1 | - | 2 - 3 | 321 | 88 | ||
DQ812116.1 | - | 2 - 3 | 186 | 88 | ||
EB956321.1 | - | 2 - 3 | 376 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
