Assay Details
Gene Symbol: tnfrsfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131840.2 | NP_571915.2 | 6 - 7 | 874 | 144 | |
GenBank mRNA | AF250042.2 | - | 6 - 7 | 804 | 144 | |
BC162652.1 | - | 6 - 7 | 774 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
