Assay Details
Gene Symbol: cotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200012.1 | NP_956306.1 | 2 - 3 | 288 | 80 | |
GenBank mRNA | AY398329.1 | - | 2 - 3 | 339 | 80 | |
BC057423.1 | - | 2 - 3 | 288 | 80 | ||
BC127564.1 | - | 2 - 3 | 259 | 80 | ||
BC164236.1 | - | 2 - 3 | 200 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
