Assay Details
Gene Symbol: mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328585.1 | NP_001315514.1 | 4 - 5 | 532 | 106 | |
NM_200307.2 | NP_956601.2 | 4 - 5 | 419 | 106 | ||
GenBank mRNA | BC050166.1 | - | 4 - 5 | 406 | 106 | |
EH546243.1 | - | 4 - 5 | 513 | 106 | ||
EH576059.1 | - | 4 - 5 | 685 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
