Assay Details
Gene Symbol: LOC108179221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017354423.1 | XP_017209912.1 | 885 | 63 |
Gene Symbol: ddx41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AI584875.1 | - | 65 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
